accessibility menu, dialog, popup

UserName

ClH3CH3CO(S)OOO(S)OHHNNHONOOH

CAS RN 144481-98-1

IUPAC Name: hydrogen [(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl 3-{4-[(2S)-2-hydroxy-3-({2-[(morpholine-4-carbonyl)amino]ethyl}amino)propoxy]phenyl}propanoate chloride
Synonyms: hydrogen landiolol chloride
Molecular Weight (g/mol): 546.06
Molecular Formula: C25H40ClN3O8
InChi Key: DLPGJHSONYLBKP-IKGOIYPNSA-N
SMILES: [H+].[Cl-].CC1(C)OC[C@@H](COC(=O)CCC2=CC=C(OC[C@@H](O)CNCCNC(=O)N3CCOCC3)C=C2)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results