missing translation for 'onlineSavingsMsg'
Learn More

Landiolol Hydrochloride 98.0+%, TCI America™

Catalog No. L029650MG
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L029650MG 50 mg
L0296250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L029650MG Supplier TCI America Supplier No. L029650MG

Chemical Identifiers

CAS 144481-98-1
Molecular Formula C25H40ClN3O8
Molecular Weight (g/mol) 546.058
MDL Number MFCD00911043
InChI Key DLPGJHSONYLBKP-IKGOIYPNSA-N
PubChem CID 46856312
IUPAC Name [(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl 3-[4-[(2S)-2-hydroxy-3-[2-(morpholine-4-carbonylamino)ethylamino]propoxy]phenyl]propanoate;hydron;chloride
SMILES [H+].CC1(OCC(O1)COC(=O)CCC2=CC=C(C=C2)OCC(CNCCNC(=O)N3CCOCC3)O)C.[Cl-]

Specifications

Melting Point 126°C
Color White
Quantity 50 mg
Formula Weight 546.06
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Landiolol Hydrochloride
RTECSNumber DA8346700
TSCA No