missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LAMP3 Monoclonal Antibody (K24013_7C5), Invitrogen™

Catalog No. MA556199
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556199 200 μL
MA555819 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556199 Supplier Invitrogen™ Supplier No. MA556199

Mouse Monoclonal Antibody

Sequence of this protein is as follows: TVSHTTGNTT QPSNQTTLPA TLSIALHKST TGQKPVQPTH APGTTAAAHN TTRTAAPAST VPGPTLAPQP SSVKTGIYQV LNGSRLCIKA EMGIQLIVQD KESVFSPRRY FNIDPNATQA SGNCGTRKSN

Dendritic cells (DCs) are the most potent antigen-presenting cells. Immature DCs efficiently capture antigens and differentiate into interdigitating dendritic cells (IDCs) in lymphoid tissues that induce primary T-cell responses (summary by de Saint-Vis et al., 1998 [PubMed 9768752]).

Specifications

Antigen LAMP3
Applications Western Blot
Classification Monoclonal
Clone K24013_7C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene LAMP3
Gene Accession No. Q9UQV4
Gene Alias 1200002D17Rik; CD208; DC LAMP; DCLAMP; DC-LAMP; DC-lysosome-associated membrane glycoprotein; dendritic cells-lysosome associated membrane glycoprotein; LAMP; Lamp3; LAMP-3; lysosomal associated membrane protein 3; lysosomal-associated membrane protein 3; lysosome-associated membrane glycoprotein 3; Protein TSC403; TSC403
Gene Symbols LAMP3
Host Species Mouse
Immunogen Recombinant human LAMP3 (immunogen range is 151-280).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27074
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less