Learn More
LaMotte™ Insta-Test™ Single and Multi-Factor Test Strips
For water quality testing
Supplier: LaMotte™ 3038 G/RSD
Description
- No reagents or field kits required for most strips
- Chemically treated pads on plastic strips produce color change
- Pop-top vials have molded desiccant liner and hinged snap cap to protect from moisture
- As little as 10-second development time
Test strips for testing water: wastewater, industrial water boiling and cooling, drinking water, laundry and sanitation †
Chemical Identifiers
| Water quality testing | |
| Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight |
Specifications
| Water quality testing | |
| 25 Strips/Factor | |
| Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight |
| NO3, NO2, pH, Alkalinity, Hardness | |
| 20-160, 0-10, 6-9, 0-240, 0-180 ppm | |
| Multi-Factor Test Strip |