missing translation for 'onlineSavingsMsg'
Learn More

LaMotte™ Insta-Test™ Single and Multi-Factor Test Strips

Catalog No. p-3256647
Change view
Click to view available options
Parameter Specifications:
Alkalinity
Ammonia
Chlorine - free, high range
Chlorine - free, low range
Chlorine - Total
Chlorine - total, low range
Chlorine dioxide
Copper
Hardness
Hydrogen Peroxide
Iron and Copper
Nitrate and Nitrite
NO3, NO2, pH, Alkalinity, Hardness
Peracetic acid
pH - Wide Range
Sodium Chloride
Total Chlorine and pH
Quantity:
10/Blister Card, 10 Cards/Pk.
25 Strips/Factor
25 Strips/Vial
25 Tests/Factor/Vial
25 Tests/Vial; 12 Vials/Cs.
50 Strips/Vial
Range:
0 to 10 ppm
0 to 180 ppm
0 to 50 ppm
0 to 500 ppm
0 to 6 ppm
0-0.3-0.6-1-3 ppm
0-0.5-1-3-5 ppm
0-0.5-1-3-5-10 ppm
0-10-20-40-60-85-160 ppm
0-5 ppm (Fe), 0-3 ppm (Cu)
0-5-10-25-50, 0-0.5-1-5-10
0-50-100-250-500-800 ppm
1500 to 5000 ppm
20-160, 0-10, 6-9, 0-240, 0-180 ppm
4-5-6-7-8-9-10
Cl: 0-1-5-10-20-50 ppm, pH: 4-10
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Parameter Specifications Quantity Range
03920375 Chlorine - free, high range 50 Strips/Vial 0-50-100-250-500-800 ppm
22307874 Peracetic acid 50 Strips/Vial 0-10-20-40-60-85-160 ppm
22307872 Chlorine dioxide 50 Strips/Vial 0 to 500 ppm
22307873 Peracetic acid 50 Strips/Vial 0 to 50 ppm
22307871 Chlorine dioxide 50 Strips/Vial 0 to 10 ppm
22307876 Hydrogen Peroxide 25 Strips/Vial 0 to 50 ppm
03920400 Chlorine - free, low range 25 Strips/Vial 0-0.5-1-3-5-10 ppm
03920401 Chlorine - total, low range 25 Strips/Vial 0 to 10 ppm
03920402 Copper 25 Strips/Vial 0-0.3-0.6-1-3 ppm
03920404 Sodium Chloride 10/Blister Card, 10 Cards/Pk. 1500 to 5000 ppm
03920399 Hardness 50 Strips/Vial 0 to 180 ppm
03920398 Nitrate and Nitrite 50 Strips/Vial 0-5-10-25-50, 0-0.5-1-5-10
03920403 Iron and Copper 25 Tests/Factor/Vial 0-5 ppm (Fe), 0-3 ppm (Cu)
03920420 Ammonia 25 Strips/Vial 0 to 6 ppm
03920421 NO3, NO2, pH, Alkalinity, Hardness 25 Strips/Factor 20-160, 0-10, 6-9, 0-240, 0-180 ppm
03920422 Alkalinity 25 Strips/Vial 0 to 180 ppm
03920423 Chlorine - Total 50 Strips/Vial 0-0.5-1-3-5 ppm
03920425 pH - Wide Range 50 Strips/Vial 4-5-6-7-8-9-10
03920426 Total Chlorine and pH 25 Tests/Vial; 12 Vials/Cs. Cl: 0-1-5-10-20-50 ppm, pH: 4-10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Catalog No. 03920375 Supplier LaMotte™ Supplier No. 3031
Only null left

For water quality testing

  • No reagents or field kits required for most strips
  • Chemically treated pads on plastic strips produce color change
  • Pop-top vials have molded desiccant liner and hinged snap cap to protect from moisture
  • As little as 10-second development time

Test strips for testing water: wastewater, industrial water boiling and cooling, drinking water, laundry and sanitation †

†Strips have been evaluated for use in these applications. Use in other applications is subject to potential interferences.

Specifications

For Use With (Application) Water quality testing
Parameter Specifications Chlorine - free, high range
Quantity 50 Strips/Vial
Range 0-50-100-250-500-800 ppm
Storage Requirements Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight
Type Single Factor Test Strip