missing translation for 'onlineSavingsMsg'
Learn More

Lamotrigine, 98%

CAS: 84057-84-1 | C9H7Cl2N5 | 256.09 g/mol

$367.45 - $367.45

Chemical Identifiers

CAS 84057-84-1
Molecular Formula C9H7Cl2N5
Molecular Weight (g/mol) 256.09
MDL Number MFCD00865333
InChI Key PYZRQGJRPPTADH-UHFFFAOYSA-N
Synonym lamotrigine, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine, lamictal, lamictal cd, lamotrigina, lamotriginum, lamictal xr, lamotriginum latin, lamotrigina spanish, lamictal odt
PubChem CID 3878
ChEBI CHEBI:6367
IUPAC Name 6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine
SMILES NC1=NN=C(C(N)=N1)C1=CC=CC(Cl)=C1Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC460850050
View Documents
Thermo Scientific Chemicals
460850050
5 g
Each for $367.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

84057-84-1
256.09
PYZRQGJRPPTADH-UHFFFAOYSA-N
3878
6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine
C9H7Cl2N5
MFCD00865333
lamotrigine, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine, lamictal, lamictal cd, lamotrigina, lamotriginum, lamictal xr, lamotriginum latin, lamotrigina spanish, lamictal odt
CHEBI:6367
NC1=NN=C(C(N)=N1)C1=CC=CC(Cl)=C1Cl

Specifications

84057-84-1
0.5% max. (105°C, 3 h)
97.5% min. (HPLC)
MFCD00865333
lamotrigine, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine, lamictal, lamictal cd, lamotrigina, lamotriginum, lamictal xr, lamotriginum latin, lamotrigina spanish, lamictal odt
PYZRQGJRPPTADH-UHFFFAOYSA-N
6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine
3878
256.09
Lamotrigine
218.0°C
Authentic
C9H7Cl2N5
5 g
Solubility in water: insoluble. Other solubilities: soluble in dmf
NC1=NN=C(C(N)=N1)C1=CC=CC(Cl)=C1Cl
256.09
CHEBI:6367
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 281-901-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only