missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Laminin gamma-3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150285
Description
Laminin gamma-3 Polyclonal Antibody for IHC (P), Western Blot
Laminin is a non-collagenous protein of basement membranes. By rotary shadowing it has a cross-like shape, consisting of one long arm, (400 kD) and three short arms (200 kD). Antibodies to laminin localize exclusively in basement membranes. Laminin is found throughout the basement membrane, but the reaction with the antibodies to laminin is more intense in the lamina lucida, the part of the basement membrane adjacent to the cell membrane. Laminin binds to type IV collagen, a component exclusively localized in the basement membrane. Laminin has been shown to play a role in cell adhesion and attachment in vivo and in vitro.Specifications
| Laminin gamma-3 | |
| Polyclonal | |
| Unconjugated | |
| LAMC3 | |
| AI562206; AW240805; Lamc3; laminin gamma 3; Laminin gamma3; laminin subunit gamma 3; laminin subunit gamma-3; laminin, gamma 3; laminin-12 subunit gamma; Laminin-14 subunit gamma; laminin-15 subunit gamma; OCCM | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 23928 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9R0B6 | |
| LAMC3 | |
| Recombinant Laminin Gamma 3 (LAMC3) (Gly751-Cys875). | |
| 200 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |