missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Naphthol Blue Disodium Salt, Reagent, ACS,, Spectrum™ Chemical

Catalog No. 18603289
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603289 100 g Amber Glass Bottle
18603288 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18603289 Supplier Spectrum Chemical Mfg Cor Supplier No. H1092100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

H1092, 165660-27-5, C20H12O11N2S3Na2

Spectrum™ Chemical Hydroxy Naphthol Blue Disodium Salt, Reagent, ACS

Chemical Identifiers

CAS 165660-27-5
Molecular Formula C20H14N2Na2O11S3
Molecular Weight (g/mol) 600.49
InChI Key LXJHFXHZWYBBKJ-UHFFFAOYSA-N
IUPAC Name 3-hydroxy-4-[2-(2-oxo-4-sulfo-1,2-dihydronaphthalen-1-ylidene)hydrazin-1-yl]naphthalene-2,7-disulfonic acid disodium
SMILES [Na].[Na].OC1=C(NN=C2C(=O)C=C(C3=CC=CC=C23)S(O)(=O)=O)C2=CC=C(C=C2C=C1S(O)(=O)=O)S(O)(=O)=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Color Blue/Purple
Quantity 100 g
Grade ACS
Chemical Name or Material Hydroxy Naphthol Blue Disodium Salt