missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Laminin alpha-5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149838
Description
Laminin alpha-5 Polyclonal Antibody for Western Blot, ICC/IF, IHC
Components of the extracellular matrix exert myriad effects on tissues throughout the body. In particular, the laminins, a family of heterotrimeric extracellular glycoproteins, affect tissue development and integrity in such diverse organs as the kidney, lung, skin, and nervous system. It is thought that laminins mediate the attachment, migration, and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. Laminins function as heterotrimeric complexes of alpha, beta, and gamma chains, with each chain type representing a different subfamily of proteins. The protein encoded by this gene belongs to the alpha subfamily of laminin chains and is a major component of basement membranes. Two transcript variants encoding different isoforms have been found for this gene, but the full-length nature of one of them has not been determined.Specifications
| Laminin alpha-5 | |
| Polyclonal | |
| Unconjugated | |
| LAMA5 | |
| [a]5; AA408760; AA408762; AI853660; KIAA0533; KIAA1907; LAMA5; laminin alpha-5 chain; laminin subunit alpha 5; laminin subunit alpha-5; laminin, alpha 5; laminin-10 subunit alpha; Laminin-11 subunit alpha; laminin-15 subunit alpha; laminin-511; mKIAA0533; RP11-157P1.6 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 16776 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q61001 | |
| LAMA5 | |
| Recombinant Laminin Alpha 5 (LAMa5) (Glu3517-His3718). | |
| 200 μL | |
| Primary | |
| Mouse | |
| Antibody |