missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Laminin alpha-1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149778
Description
Laminin alpha-1 Polyclonal Antibody for Western Blot, ICC/IF, IHC
Laminin antibodies are widely used to label blood vessels and basement membranes.Spécifications
| Laminin alpha-1 | |
| Polyclonal | |
| Unconjugated | |
| LAMA1 | |
| LAMA; LAMA1; Laminin A chain; laminin subunit alpha 1; laminin subunit alpha-1; laminin, alpha 1; Laminin-1 subunit alpha; Laminin-3 subunit alpha; PTBHS; S-LAM alpha; S-LAM-alpha; S-laminin subunit alpha | |
| Rabbit | |
| Protein A | |
| RUO | |
| 284217 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P25391 | |
| LAMA1 | |
| Recombinant Laminin Alpha 1 (LAMA1) (Ala886-Val1039). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |