missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lamin B Receptor Recombinant Rabbit Monoclonal Antibody (23GB6100)

Catalog No. MA552985
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552985 20 μL
MA552984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552985 Supplier Invitrogen™ Supplier No. MA552985
Only null left

Rabbit Recombinant Monoclonal Antibody

Lamin B Receptor Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

The protein encoded by this gene belongs to the ERG4/ERG24 family. It localized in the nuclear envelope inner membrane and anchors the lamina and the heterochromatin to the membrane. It may mediate interaction between chromatin and lamin B. Mutations of this gene has been associated with autosomal recessive HEM/Greenberg skeletal dysplasia. Alternative splicing occurs at this locus and two transcript variants encoding the same protein have been identified.

Specifications

Antigen Lamin B Receptor
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB6100
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LBR
Gene Accession No. O08984, Q14739
Gene Alias 3-beta-hydroxysterol Delta (14)-reductase; AI505894; C-14 sterol reductase; C14SR; Delta(14)-sterol reductase LBR; delta(14)-sterol reductase LBR; lamin-B receptor; Delta-14-SR; DHCR14B; FLJ43126; ic; ichthyosis; integral nuclear envelope inner membrane protein; lamin B receptor; lamin-B receptor; lamin-b receptor-like protein; Lbr; LMN2R; MGC9041; NBP60; PHA; PRO0650; sb:cb406; Sterol C14-reductase; TDRD18; tudor domain containing 18; wu:fb75h08; wu:fc47b04; wu:fd36b07; zgc:86649
Gene Symbols LBR
Host Species Rabbit
Immunogen A synthesized peptide derived from human Lamin B Receptor (300-615AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3930, 89789
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less