missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lamin A/C Monoclonal Antibody (Jol3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Fisher Scientific MA15820
Description
MA1-5820 reacts with human dermal fibroblasts in immunocytochemistry. Other cell/tissue types have not been tested in this application. It reacts with human, mouse and rat samples in Western blot. This antibody recognizes the common tail domain of Lamin A+C, epitope aa 464-572.
Lamins are a class of intermediate filament proteins that form a matrix on the inner surface of the nuclear envelope. These proteins are found in many different cell types in three different forms (A, B, and C). Lamins A and C are alternatively spliced versions of the LMNA gene. The LMNA gene has been linked to many disorders of the muscular system, nervous system, and the fat distributions systems including: Emery-Dreifuss muscular dystrophy, Dunnigan-type familial partial lipodystrophy (FPLD), limb-girdle muscular dystrophy (LGMD1B), dilated cardiomyopathy (CMD1A), axonal neuropathy (Charcot-Marie-Tooth disease; CMT2B1), and mandibuloacral dysplasia (MAD).Chemical Identifiers
| Lamin A/C | |
| Conc. Not Determined | |
| Monoclonal | |
| Liquid | |
| RUO | |
| tissue culture supernatant with 0.1% sodium azide | |
| 70 kDa lamin; cb948; CDCD1; CDDC; CMD1A; CMT2B1; Dhe; EMD2; FPL; FPLD; FPLD2; HGPS; I79_009616; IDC; lamin; lamin A; lamin A/C; lamin A/C L homeolog; lamin A/C-like 1; Lamin A+C mutant; Lamin AC; lamin a-c; lamin C; lamin C2; lamin-A; Lamin-A/C; LDP1; LFP; LGMD1B; LMN1; lmna; LMNA protein; lmna.L; lmna-A; LMNC; LMNL1; mutant 453W; Mutant lamin A/C; nuclear intermediate filament protein; nuclear lamin A; prelamin-A/C; PRO1; progerin mutant; Renal carcinoma antigen NY-REN-32; RP11-54H19.1; unnamed protein product; wu:fk66d12; XELAEV_18040808mg | |
| LMNA | |
| Primary | |
| Antibody |
| Jol3 | |
| ChIP Assay, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| P02545, P48678, P48679 | |
| 16905, 4000, 60374 | |
| IgG1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| LMNA |
Specifications
| Lamin A/C | |
| Monoclonal | |
| Conc. Not Determined | |
| tissue culture supernatant with 0.1% sodium azide | |
| P02545, P48678, P48679 | |
| LMNA | |
| Recombinant bacterially expressed full length protein | |
| RUO | |
| 16905, 4000, 60374 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunocytochemistry, Western Blot | |
| Jol3 | |
| Unconjugated | |
| LMNA | |
| 70 kDa lamin; cb948; CDCD1; CDDC; CMD1A; CMT2B1; Dhe; EMD2; FPL; FPLD; FPLD2; HGPS; I79_009616; IDC; lamin; lamin A; lamin A/C; lamin A/C L homeolog; lamin A/C-like 1; Lamin A+C mutant; Lamin AC; lamin a-c; lamin C; lamin C2; lamin-A; Lamin-A/C; LDP1; LFP; LGMD1B; LMN1; lmna; LMNA protein; lmna.L; lmna-A; LMNC; LMNL1; mutant 453W; Mutant lamin A/C; nuclear intermediate filament protein; nuclear lamin A; prelamin-A/C; PRO1; progerin mutant; Renal carcinoma antigen NY-REN-32; RP11-54H19.1; unnamed protein product; wu:fk66d12; XELAEV_18040808mg | |
| Mouse | |
| 1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |