missing translation for 'onlineSavingsMsg'
Learn More
Learn More
α-Lactose, MilliporeSigma™ Supelco™
Description
Featured Industries: Food and Beverages
Specifications
Specifications
| Packaging | Ampule |
| CAS | 5989-81-1 |
| Physical Form | Neat |
| Linear Formula | C12H22O11 · H2O |
| MDL Number | MFCD00150747 |
| Synonym | alpha-Lactose monohydrate; beta-D-Gal-(1->4)-alpha-D-Glc; 4-O-beta-D-Galactopyranosyl-alpha-D-glucose; Milk sugar |
| Type | Analytical Standard |
| Quantity | 500 mg |
| Formula Weight | 360.31 |
| Grade | Analytical Standard |