missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid, Certified, 1.00N ±0.05N (1.0M), LabChem™

Catalog No. p-5388954
Change view
Click to view available options
Quantity:
500 mL
1 L
Catalog No. Quantity
LC160201 500 mL
LC160202 1 L
2 options

Catalog No. LC160201

Supplier: LabChem LC160201

Only null left

Chemical Identifiers

CAS 50-21-5 , 7732-18-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
PubChem CID 612
ChEBI CHEBI:78320
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

Color Colorless
CAS Max % 91.15, 8.85
Linear Formula C3H6O3
Solubility Information Soluble in water
Concentration 1.00N ±0.05N (1.0M)
Formula Weight 90.08
Grade Certified
Density 1.02g/mL
Quantity 500 mL
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.02g/mL
Traceability to NIST Traceable to NIST
Chemical Name or Material Lactic Acid
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature