missing translation for 'onlineSavingsMsg'
Learn More

3M™ Petrifilm™ Lactic Acid Bacteria Count Plate

Numéro de catalogue. S30733
Modifier l'affichage
Click to view available options
Quantité:
50 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
S30733 50 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. S30733 Fournisseur 3M™ Petrifilm™ Code fournisseur 6461

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Quickly and reliably detect lactic acid bacteria with sample-ready count plates. No prep, gas packs, or CO2 chambers required.

  • Product Type: Count Plate
  • Test Type: Environmental and Food Sample Test
  • Application: Detects total lactic acid bacteria populations in food and environmental samples
  • Incubation Time: Results in 48 hours
  • Format: Sample-ready plates – no media preparation needed
  • Anaerobic Testing: Built-in oxygen-scavenging and barrier films create self-contained anaerobic environment
  • No Additional Equipment Needed: Eliminates the need for gas packs, chambers, or CO2 incubators
  • PTM #041701 for selected foods
  • NF VALIDATION 3M 01/19-11/17 by AFNOR Certification (excluding yogurts)
  • Versatility: Compatible with a broad range of sample types and proven against MRS/APT agar methods
  • Pack Size: 500 plates

Spécifications

Détecte Total lactic acid bacteria populations
Type de produit Count Plate
Type d’essai Environmental Test
À utiliser avec (application) Food and environmental samples
Quantité 50 Pack