missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid 85%, Purified, Reagents

Catalog No. p-200021229
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337588 100 mL
01337587 4 L
01337586 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337588 Supplier Labchem Inc Supplier No. C2108000100A
Only null left

Lactic Acid 85%, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 50-21-5 , 7732-18-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym 2-Hydroxypropanoic Acid, 2-hydroxypropionic Acid
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

pH 1.75
Melting Point 16.8°C
Boiling Point 122°C
Color Colorless to Slight Yellow
Grade Pure
Quantity 100 mL
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.206
Chemical Name or Material Lactic Acid 85%, Purified