missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid 85%, Purified, Reagents

Catalog No. 01337588
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337588 100 mL
01337586 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337588 Supplier Labchem Inc Supplier No. C2108000100A
Only null left

Lactic Acid 85%, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 50-21-5 , 7732-18-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym 2-Hydroxypropanoic Acid, 2-hydroxypropionic Acid
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

pH 1.75
Melting Point 16.8°C
Boiling Point 122°C
Color Colorless to Slight Yellow
Grade Pure
Quantity 100 mL
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.206
Chemical Name or Material Lactic Acid 85%, Purified