missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactic Acid 85%, Purified, Reagents
Lactic Acid 85%, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C21080004A
Specifications
| Lactic Acid 85%, Purified | |
| 16.8°C | |
| 122°C | |
| C3H6O3 | |
| JVTAAEKCZFNVCJ-UHFFFAOYNA-N | |
| 2-hydroxypropanoic acid | |
| Pure | |
| Poly Bottle | |
| 1.206 |
| 1.75 | |
| 50-21-5,7732-18-5 | |
| Colorless to Slight Yellow | |
| 2-Hydroxypropanoic Acid, 2-hydroxypropionic Acid | |
| CC(O)C(O)=O | |
| 90.08 | |
| 4 L | |
| Liquid |
Chemical Identifiers
| 50-21-5 | |
| 90.08 | |
| 2-Hydroxypropanoic Acid, 2-hydroxypropionic Acid | |
| CC(O)C(O)=O |
| C3H6O3 | |
| JVTAAEKCZFNVCJ-UHFFFAOYNA-N | |
| 2-hydroxypropanoic acid |