missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid 85%, ACS Reagent, Reagents

Catalog No. p-200021088
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338174 4 L
01338173 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01338174 Supplier Labchem Inc Supplier No. C11080004A
Only null left

Lactic Acid 85%, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 50-21-5 , 7732-18-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym 2-Hydroxypropanoic Acid, 2-hydroxypropionic Acid
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

pH 1.75
Melting Point 16.8°C
Boiling Point 122°C
Color Colorless to Slightly Yellow
Grade ACS
Quantity 4 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.206
Chemical Name or Material Lactic Acid 85%, ACS Reagent