missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactic Acid 85%, ACS Reagent, Reagents
Specifications
Specifications
| pH | 1.75 |
| Melting Point | 16.8°C |
| Boiling Point | 122°C |
| Color | Colorless to Slightly Yellow |
| Quantity | 4 L |
| Packaging | Poly Bottle |
| Physical Form | Liquid |
| Specific Gravity | 1.206 |
| Grade | ACS |
| Chemical Name or Material | Lactic Acid 85%, ACS Reagent |