missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid, 85%, ACS Reagent Grade, Ricca Chemical

Supplier:  Ricca Chemical Company RABL0010500A

Catalog No. RABL0010500


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Lactic Acid
Clear colorless liquid
dihydrogen oxide, dihydrogen monoxide
90.08 g/mol
ACS
Odorless to slightly acrid
Solution
50-21-5 , 7732-18-5
85%
Solubility in Water: Infinite
85% w/w
500 mL
Natural Poly Bottle

Chemical Identifiers

50-21-5
90.08 g/mol
dihydrogen oxide, dihydrogen monoxide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Corrosive

SDS Documents

Documents

Promotions

Promotions