missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid, 20% (w/v), Ricca Chemical

Catalog No. p-3251846
Change view
Click to view available options
Quantity:
500 mL
10 L
Packaging:
Cubitainer
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
427216 500 mL Natural Poly Bottle
427225 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 427216 Supplier Ricca Chemical Company Supplier No. 427216

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50-21-5 , 7732-18-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
PubChem CID 612
ChEBI CHEBI:78320
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

CAS Min % 80.0, 18.2
CAS Max % 83.0, 18.94
Quantity 500 mL
Packaging Natural Poly Bottle
Chemical Name or Material Lactic Acid