missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid, 10% (v/v), Ricca Chemical

Catalog No. p-2916698
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Catalog No. Quantity Packaging
427016 500 mL Natural Poly Bottle
1 options

Catalog No. 427016

Supplier: Ricca Chemical Company 427016

Only null left

Chemical Identifiers

CAS 7732-18-5 , 50-21-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) Mixture
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
PubChem CID 612
ChEBI CHEBI:78320
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

CAS Min % 89.0, 9.0
CAS Max % 91.0, 11.0
Percent Purity 10% v/v
Grade Laboratory
Concentration or Composition (by Analyte or Components) 10% (v/v) Aqueous Solution
Quantity 500 mL
Packaging Natural Poly Bottle
Chemical Name or Material Lactic Acid