missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid, 1% v/v, Reagents

Catalog No. p-200021819
Change view
Click to view available options
Quantity:
4 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334846 10 L
01334845 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334846 Supplier Labchem Inc Supplier No. CL10130010F
Only null left

Lactic Acid, 1% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 50-21-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym 2-Hydroxypropanoic Acid, 2-hydroxypropionic Acid
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
Percent Purity 1%
Grade Reagent
Quantity 10 L
Packaging Cubitainer
Physical Form Liquid
Specific Gravity 1.0021
Chemical Name or Material Lactic Acid, 1% v/v