missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactic Acid, 1% v/v, Reagents
Specifications
Specifications
| Boiling Point | 100°C |
| Color | Colorless |
| Quantity | 10 L |
| Packaging | Cubitainer |
| Physical Form | Liquid |
| Specific Gravity | 1.0021 |
| Percent Purity | 1% |
| Grade | Reagent |
| Chemical Name or Material | Lactic Acid, 1% v/v |