missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid, 1.00 N, Ricca Chemical

$171.97 - $240.20

Chemical Identifiers

CAS 7732-18-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) Mixture
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
PubChem CID 612
ChEBI CHEBI:78320
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
4280-16
View Documents
Ricca Chemical Company
428016
500 mL Natural Poly Bottle
Each for $171.97
Only null left
 
428032
View Documents
Ricca Chemical Company
428032
1 L Natural Poly Bottle
Each for $240.20
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
Mixture
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
CHEBI:78320
CC(O)C(O)=O
C3H6O3
MFCD00004520
612
2-hydroxypropanoic acid

Specifications

7732-18-5,50-21-5
Laboratory
500 mL
Lactic Acid
dihydrogen oxide, dihydrogen monoxide
1.00N
Natural Poly Bottle
Videos
SDS
Documents

Documents