missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid, 0.100 Normal, Ricca Chemical

Catalog No. 427516
Change view
Click to view available options
Quantity:
500mL
Packaging:
Poly natural
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
427516 500mL Poly natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 427516 Supplier Ricca Chemical Company Supplier No. RI427516

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50-21-5 , 7732-18-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
PubChem CID 612
ChEBI CHEBI:78320
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

CAS Min % 97.0, 0.99
CAS Max % 100.0, 1.04
Quantity 500mL
Packaging Poly natural
Chemical Name or Material Lactic Acid