missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactamide 98.0+%, TCI America™
$178.95 - $328.08
Chemical Identifiers
| CAS | 2043-43-8 |
|---|---|
| Molecular Formula | C3H7NO2 |
| Molecular Weight (g/mol) | 89.094 |
| MDL Number | MFCD00008017 |
| InChI Key | SXQFCVDSOLSHOQ-UHFFFAOYSA-N |
| Synonym | 2-Hydroxypropionamide |
| PubChem CID | 94220 |
| ChEBI | CHEBI:75144 |
| IUPAC Name | 2-hydroxypropanamide |
| SMILES | CC(C(=O)N)O |
Chemical Identifiers
| 2043-43-8 | |
| 89.094 | |
| SXQFCVDSOLSHOQ-UHFFFAOYSA-N | |
| 94220 | |
| 2-hydroxypropanamide |
| C3H7NO2 | |
| MFCD00008017 | |
| 2-Hydroxypropionamide | |
| CHEBI:75144 | |
| CC(C(=O)N)O |
Specifications
| 2043-43-8 | |
| White-Yellow | |
| MFCD00008017 | |
| 2-Hydroxypropionamide | |
| CC(C(=O)N)O | |
| 89.094 | |
| CHEBI:75144 | |
| ≥98.0% (N) | |
| Lactamide |
| 78°C | |
| C3H7NO2 | |
| 10 g | |
| SXQFCVDSOLSHOQ-UHFFFAOYSA-N | |
| 2-hydroxypropanamide | |
| 94220 | |
| 89.09 | |
| Crystalline Powder |
Safety and Handling
TSCA : No