missing translation for 'onlineSavingsMsg'
Learn More

Lactamide 98.0+%, TCI America™

Catalog No. L000110G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L000110G 10 g
L000125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L000110G Supplier TCI America Supplier No. L000110G
Only null left

Chemical Identifiers

CAS 2043-43-8
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.094
MDL Number MFCD00008017
InChI Key SXQFCVDSOLSHOQ-UHFFFAOYSA-N
Synonym 2-Hydroxypropionamide
PubChem CID 94220
ChEBI CHEBI:75144
IUPAC Name 2-hydroxypropanamide
SMILES CC(C(=O)N)O

Specifications

Melting Point 78°C
Color White-Yellow
Quantity 10 g
Formula Weight 89.09
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Lactamide