missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lac beta(1-4)Lac-beta-C10 98.0+%, TCI America™
Supplier: TCI America L022910MG
Specifications
| Lac beta(1-4)Lac-beta-C10 | |
| White | |
| 10 mg | |
| OABQMJAOTJPXDB-XQKMEKMUSA-N | |
| (2S,3R,4S,5R,6R)-2-[(2R,3S,4R,5R,6S)-6-[(2R,3R,4R,5R,6S)-6-[(2R,3S,4R,5R,6R)-6-decoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol | |
| 121235355 | |
| ≥98.0% (HPLC) |
| 1858224-00-6 | |
| C34H62O21 | |
| Decyl 4-O-beta-D-Lactopyranosyl-beta-D-lactopyranoside | |
| CCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)OC3C(C(C(C(O3)CO)OC4C(C(C(C(O4)CO)O)O)O)O)O)O)O)O)O | |
| 806.849 | |
| 806.85 | |
| Crystalline Powder |
Chemical Identifiers
| 1858224-00-6 | |
| 806.849 | |
| Decyl 4-O-beta-D-Lactopyranosyl-beta-D-lactopyranoside | |
| (2S,3R,4S,5R,6R)-2-[(2R,3S,4R,5R,6S)-6-[(2R,3R,4R,5R,6S)-6-[(2R,3S,4R,5R,6R)-6-decoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
| C34H62O21 | |
| OABQMJAOTJPXDB-XQKMEKMUSA-N | |
| 121235355 | |
| CCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)OC3C(C(C(C(O3)CO)OC4C(C(C(C(O4)CO)O)O)O)O)O)O)O)O)O |
Safety and Handling
TSCA : No
Recommended Storage : Freezer