accessibility menu, dialog, popup

UserName

CAS RN 1858224-00-6

H3COOHOOOHOOOOHOOOHOHOHOHOHHOOHOHOHOH

CAS RN 1858224-00-6

IUPAC Name: 2-({6-[(6-{[6-(decyloxy)-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl)oxy]-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl}oxy)-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: 2-({6-[(6-{[6-(decyloxy)-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl)oxy]-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl}oxy)-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol): 806.85
Molecular Formula: C34H62O21
InChi Key: OABQMJAOTJPXDB-UHFFFAOYNA-N
SMILES: CCCCCCCCCCOC1OC(CO)C(OC2OC(CO)C(OC3OC(CO)C(OC4OC(CO)C(O)C(O)C4O)C(O)C3O)C(O)C2O)C(O)C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results