Learn More
Thermo Scientific™ Labtainer™ 2D BioProcess Container (BPC), Aegis™ 5-14 film, three ports, Luer locks

Thermo Scientific Labtainer 2D BioProcess Containers (BPCs) are composed of Aegis 5-14 single-web, multi-layer film.
$178.63 - $960.43
Specifications
| Autoclavable | Not Autoclavable |
|---|---|
| Certifications/Compliance | ISO 7 |
| Color | Transparent |
| Disposable | Single-use |
| For Use With (Application) | Bioprocessing |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
SH3105011
![]() |
Thermo Scientific™
SH3105011 |
50 mL |
Pack of 10 Each for $529.97
|
|
|||||
SH3105012
![]() |
Thermo Scientific™
SH3105012 |
100 mL |
Pack of 10 Each for $731.96
|
|
|||||
SH3105013
![]() |
Thermo Scientific™
SH3105013 |
250 mL |
Pack of 10 Each for $960.43
|
|
|||||
SH3105014
![]() |
Thermo Scientific™
SH3105014 |
500 mL |
Each of 10 for $806.98
|
|
|||||
SH3105015
![]() |
Thermo Scientific™
SH3105015 |
1 L |
Pack of 10 Each for $670.00
|
|
|||||
SH3105016
![]() |
Thermo Scientific™
SH3105016 |
2000 mL |
Pack of 10 Each for $795.91
|
|
|||||
SH3104901
![]() |
Thermo Scientific™
SH3104901 |
5 L |
Each for $178.63
|
|
|||||
SH3104902
![]() |
Thermo Scientific™
SH3104902 |
10 L |
Each for $189.08
|
|
|||||
SH3104903
![]() |
Thermo Scientific™
SH3104903 |
20 L |
Each for $199.54
|
|
|||||
SH3104904
![]() |
Thermo Scientific™
SH3104904 |
50 L |
Each for $295.67
|
|
|||||
SH3104905
![]() |
Thermo Scientific™
SH3104905 |
2 L |
Each for $191.42
|
|
|||||
Description
Features of the Labtainer 2D BPC include:
• Helps eliminate post-use cleaning steps required with re-useable containers and reduce cross-contamination risks
• Constructed in an ISO 7 cleanroom under cGMP conditions
Applications of the Labtainer 2D BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors
• Prepare and store buffer, culture media, and process liquid
• Collect chromatography feed and fractions
• Harvest, storage, and transport bulk drug product and bulk drug precursors
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands
Specifications
| Not Autoclavable | |
| Transparent | |
| Bioprocessing | |
| C-Flex | |
| 3 ports |
| ISO 7 | |
| Single-use | |
| Sterile | |
| Aegis 5-14 |