missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Labtainer™ 2D BioProcess Container (BPC), Aegis™ 5-14 film, three ports, Luer locks
GREENER_CHOICE

Catalog No. SH3105011
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1 L
2 L
2000 mL
5 L
10 L
20 L
50 L
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
SH3105011 50 mL
SH3105012 100 mL
SH3105013 250 mL
SH3105014 500 mL
SH3105015 1 L
SH3105016 2000 mL
SH3104901 5 L
SH3104902 10 L
SH3104903 20 L
SH3104904 50 L
SH3104905 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. SH3105011 Supplier Thermo Scientific™ Supplier No. SH3105011

Thermo Scientific Labtainer 2D BioProcess Containers (BPCs) are composed of Aegis 5-14 single-web, multi-layer film.

Features of the Labtainer 2D BPC include:
• Helps eliminate post-use cleaning steps required with re-useable containers and reduce cross-contamination risks
• Constructed in an ISO 7 cleanroom under cGMP conditions

Applications of the Labtainer 2D BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors
• Prepare and store buffer, culture media, and process liquid
• Collect chromatography feed and fractions
• Harvest, storage, and transport bulk drug product and bulk drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance ISO 7
Color Transparent
Disposable Single-use
For Use With (Application) Bioprocessing
Sterility Sterile
Tubing Style C-Flex
Film Description Aegis 5-14
Port Detail 3 ports
Quantity 50 mL
Capacity (Metric) 50 mL
Show More Show Less