missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laboratory Performance Check Standard For Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC LPC1100
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| K | 100μg/mL |
| P | 100μg/mL |
| SiO&subscript_2; | 100μg/mL |
| Al | 20μg/mL |
| As | 20μg/mL |
| B | 20μg/mL |
| Ba | 20μg/mL |
| Be | 20μg/mL |
| Ca | 20μg/mL |
| Cd | 20μg/mL |
| Co | 20μg/mL |
| Cr | 20μg/mL |
| Cu | 20μg/mL |
| Fe | 20μg/mL |
| Li | 20μg/mL |
| Mg | 20μg/mL |
| Mn | 20μg/mL |
| Mo | 20μg/mL |
| Na | 20μg/mL |
| Ni | 20μg/mL |
| Pb | 20μg/mL |
| Sb | 20μg/mL |
| Se | 20μg/mL |
| Sn | 20μg/mL |
| Sr | 20μg/mL |
| Tl | 20μg/mL |
| V | 20μg/mL |
| Zn | 20μg/mL |
| Ag | 5μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| LPC Standard 1 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Nitric Acid/Trace Tartaric Acid/Trace Hydrogen Fluoride | |
| Environmental |
| Triple-leached LDPE bottle | |
| AA, ICP | |
| Certified Reference Material | |
| 125 mL | |
| Various |
Product intended for laboratory use only