Learn More
Fisherbrand™ Laboratory Ball Valve
Laboratory Ball Valve is deck mounted.
Description
- Chrome plated brass
- Quarter-turn open/close
- Forged brass lever with color-coded index disc
Specifications
| Chrome | |
| Chrome-plated Brass | |
| Ball valve assembly | |
| 5.7 in. | |
| 1.6 in. | |
| 3.6 in. |
| Orange | |
| 1 Turret, Air Service | |
| 3/8 in. IPS mounting shank with a 3/8 in. NPT male inlet | |
| 14.5 cm | |
| 4.1 cm | |
| 9.2 cm |