Learn More
Scientific Device Laboratory Adhesive Coated Slides
Description
- Adhesives will not interfere with staining
- Ideal for hybridization techniques
- Frosted ends
Specifications
Specifications
| Frosted | 1 End |
| Coating | Poly-L-Lysine |
| Quantity | 720/Cs. |