missing translation for 'onlineSavingsMsg'
Learn More

STERIS™ LabKlenz 120 Alkaline Detergent

LabKlenz 120 Mild Alkaline Detergent is a mild alkaline liquid detergent. It is compatible with a variety of substrates including stainless steel, mild steel, copper and aluminum. This product is extremely free rinsing to meet critical laboratory requirements. pH (1%): 9.4, typical.

$3655.77 - $3655.77

Specifications

Product Type Detergent
Color-coded No
Form Clear Liquid
pH 9.4
Scent Unscented
View More Specs

Products 1
Catalog Number Mfr. No. Size Container Type Price Quantity  
Catalog Number Mfr. No. Size Container Type Price Quantity  
17-467-595
View Documents
STERIS™
1L1202
30 gal. Drum
Each for $3,655.77
Only null left
 
Description

Description

LabKlenz 120 Detergent is specially formulated to meet the unique cleaning and materials compatibility requirements found in laboratory glassware washing applications.

HOW LABKLENZ 120 DETERGENT WORKS

LabKlenz 120 Detergent is a phosphate-free, liquid, alkaline detergent for use in automated laboratory glassware washing applications. It is compatible with a variety of substrates including stainless steel, mild steel, and copper. It is extremely free rinsing to meet critical laboratory requirements.

WHY LABKLENZ 120 DETERGENT?

  • Efficacy - Effective removal of laboratory residues in both soft and hard water
  • Flexibility - Mild alkalinity for better materials compatibility
  • Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally Friendly - phosphate-free formulation with biodegradable surfactants
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility

Physical Properties

pH (1%): 9.4, typical

Specifications

Specifications

Detergent
Clear Liquid
Unscented
No
9.4
Non-sterile
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'