missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mirus Bio™ Label IT™ Nucleic Acid Labeling Kit, MFP488
Description
- Label Any DNA or RNA Template – Suitable for a wide range of applications
- One-step Chemical Method – Easily and consistently control the labeling reactions
- Adjustable Labeling Density – Achieve high sensitivity with optimally labeled DNA and RNA
- Covalent Mechanism – Permanent, non-destructive modification of nucleic acid residues is ideal for many diverse applications; labels do not impact hybridization performance
NOTE: All the Label IT Reagents use the same technology, but each attach different fluorophores with distinct excitation and emission spectra.
- MFP488 Nucleic Acid Labeling Reagent
- High Photostability – Facilitates longer time for observation and image capture
- Widespread Instrument Compatibility – Spectrally similar to fluorescein
- Stable at Low pH – Fluorescence is maintained over a broad range (pH 4-9)
Specifications
Specifications
| Product Type | Label Reagent |
| Concentration | 1X |
| Product Line | Molecular Probes, Click-iT |
| For Use With (Application) | Labels 25 μg nucleic acid |