missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mirus Bio Label IT™ Nucleic Acid Labeling Kit, CX-Rhodamine
Label IT™ Nucleic Acid Labeling Reagents are ideal for efficient, direct, non-enzymatic labeling of DNA and RNA.
Supplier: Mirus Bio MIR3100
Description
- Nucleic Acid Labeling Reagents
- Label Any DNA or RNA Template: Suitable for a wide range of applications
- One-step Chemical Method: Easily and consistently control the labeling reactions
- Adjustable Labeling Density: Achieve high sensitivity with optimally labeled DNA and RNA
- Covalent Mechanism: Permanent, non-destructive modification of nucleic acid residues is ideal for many diverse applications; labels do not impact hybridization performance
- NOTE: All the Label IT™ Reagents use the same technology, but each attach different fluorophores with distinct excitation and emission spectra.
- MFP488 Nucleic Acid Labeling Reagent
- High Photostability: Facilitates longer time for observation and image capture
- Widespread Instrument Compatibility: Spectrally similar to fluorescein
- Stable at Low pH: Fluorescence is maintained over a broad range (pH 4-9)
Specifications
| Fluorescence | |
| Kit | |
| DNA (Labeled), RNA (Labeled) | |
| DNA/RNA |