Learn More
Labconco™ Thimble Exhaust Connectors for Precise™ Glove Boxes
Catalog No. 16108406
Change view
Click to view available options
Diameter (English):
5 in.
6 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English)
16108406 6 in.
16108427 5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16108406 Supplier Labconco™ Supplier No. LCN5230200
Accessories for use with Precise Glove Boxes
Epoxy-coated steel for connecting the glove box to ductwork attached to building exhaust, remote blower or FilterMate Portable Exhauster. 6 in. diameter.
Specifications
For Use With (Equipment) Precise HEPA-filtered Glove Box
Weight (English) 5 lb.
Weight (Metric) 2.3 Kg
Diameter (English) 6 in.
Material Epoxy-coated Steel
For Use With (Application) Connecting the glove box to building exhaust or remote blower for outside ducting