Learn More
Labconco™ Starter Kits for Forensic Enclosures
Kits include accessories needed for any type of forensic drying.
Supplier:  Labconco™ 3869910
 View more versions of this product
Catalog No. 10359254
Only null left
Description
Description
Starter Kits for Labconco Protector Evidence Drying Cabinet. Each Kit includes six Inlet Door Prefilters, six Exhaust Cabinet Prefilters, 10 Hanging Clips, 10 Security Tags, 10 Paper Liners, one 32 ounce (946 ml) bottle of Germicidal Cleaner.
Specifications
Chemical Identifiers
15 lb.
6.8 Kg
Enclosure Kit
Specifications
4 ft. Evidence Drying Cabinet
6.8 Kg
4 ft. Protector™ evidence drying cabinet
15 lb.
Enclosure Kit
Product Suggestions
Product Suggestions