missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Stainless Steel Perforated Trays for Forensic Enclosures

Catalog No. 09681286
Change view
Click to view available options
For Use With (Equipment):
Capture Portable Fuming System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
09681286 Capture Portable Fuming System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09681286 Supplier Labconco™ Supplier No. 3426101

Accessories for use with CApture Portable Fuming System.

May be placed on the Stainless Steel Shelf to support small items of evidence, such as bullet casings. Perforations allow for airflow.

Specifications

For Use With (Equipment) Capture Portable Fuming System
Weight (English) 1 lb.
Weight (Metric) 0.5 Kg
Depth (English) 5.25 in.
Width (English) 8 in.
Depth (Metric) 13.33 cm
Width (Metric) 20.3 cm
Product Type Cyanoacrylate Fuming Shelf
Material Stainless Steel
For Use With (Application) Cyanoacrylate Fuming