missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ SpillStopper™ Work Surface for 6ft. Protector™XStream

Labconco SpillStopper™ Work Surface is dished and contoured to conform to the interior liner to contain spills.

Supplier:  Labconco™ 9503611

 View more versions of this product

Catalog No. 10369194


Only null left

Description

Description

  • Dished and contoured to conform to interior liner of Protector Laboratory Hoods
  • Front edge with large radius to aerodynamically direct airflow in hood
  • Pre-drilled holes for venting and plumbing passthroughs
  • Includes left rear cupsink cutout
Specifications

Chemical Identifiers

Protector XStream Fume Hood (72 in.)
Work Surface
Epoxy Resin
Laboratory Fume Hood Work

Specifications

Protector XStream Fume Hood (72 in.)
Work Surface
Laboratory Fume Hood Work
Left Rear, Right Rear
Epoxy Resin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions