missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ SpillStopper™ Work Surfaces for 4ft. Protector™XStream

SpillStopper work surfaces are dished to contain spills

Supplier:  Labconco™ 9503411

 View more versions of this product

Catalog No. 10-369-180

Only null left
Explore available promotions

Description

Description

  • Front edge has large radius to aerodynamically direct airflow in the hood
  • Some surfaces have cutouts for a surface-mounted service fixture and/or a 3 x 6 in. cup sink
  • Flat work surfaces may also be coupled with an adjustable height base stand to provide auxiliary workspace
  • Service fixture and cup sink are sold separately
Specifications

Specifications

Protector XStream Fume Hood (48 in.)
Work Surface
Laboratory Fume Hood Work
Left Rear, Right Rear
Epoxy Resin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions