missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ SpillStopper™ Work Surfaces for 4ft. Protector™XStream

Catalog No. p-4517895
Change view
Click to view available options
Includes:
Left Rear
Left Rear, Right Rear
Left Side
Left Side, Right Rear
No Cutouts
Right Rear
Right Side
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
10369177 Right Rear
10369179 Right Side
10369176 Left Rear
10369178 Left Side
10369175 No Cutouts
10369180 Left Rear, Right Rear
10369181 Left Side, Right Rear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 10369177 Supplier Labconco™ Supplier No. 9503401
Only null left

SpillStopper work surfaces are dished to contain spills

  • Front edge has large radius to aerodynamically direct airflow in the hood
  • Some surfaces have cutouts for a surface-mounted service fixture and/or a 3 x 6 in. cup sink
  • Flat work surfaces may also be coupled with an adjustable height base stand to provide auxiliary workspace
  • Service fixture and cup sink are sold separately

Specifications

For Use With (Equipment) Protector XStream Fume Hood (48 in.)
Includes Right Rear
Product Type Work Surface
Material Epoxy Resin
For Use With (Application) Laboratory Fume Hood Work