Learn More
Labconco™ Seismic Bracket Kit
Catalog No. 10269302
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
10269302 Stainless Steel
1 options
Catalog No. 10269302 Supplier Labconco™ Supplier No. 3857000
Includes
Four 0.25 in. (0.64cm) thick stainless-steel seismic brackets and hardware for attachment to the stand. Hardware to attach to the floor and installation is required (not included).
Offers an extra measure of stability
Secures telescoping base stand with fixed-feet to the floor in earthquake-prone regions.
Specifications
Weight (English) 10 lb.
Weight (Metric) 4.53 Kg
Material Stainless Steel