Learn More
Labconco™ Secondary Trap Canister and Inserts for CentriVap™ System
Help to provide secondary protection to vacuum pumps in evaporation systems
Supplier:  Labconco™ 7815200
 View more versions of this product
Catalog No. 1631611
Only null left
Description
Description
Help to provide secondary protection to vacuum pumps in evaporation systems
  • Clear polypropylene canister with mounting bracket, 17 in. (43cm) PVC tubing, and two clamps
  • Canister accommodates trap inserts listed in the table
Specifications
Chemical Identifiers
Polypropylene
CentriVap™ system
Specifications
Polypropylene
Solvent Trap Insert
1.8 kg
CentriVap™ system
4 lb.
Product Suggestions
Product Suggestions