Learn More
Labconco™ Replacement Aluminum Heater Pans for Forensic Enclosures
Description
Disposable aluminum pans for holding Super Glue™ or water. Ten pans are included with each CApture Portable Fuming System. Package of 250.
Specifications
Specifications
| For Use With (Equipment) | Capture Portable Fuming System |
| Weight (English) | 1 lb. |
| Weight (Metric) | 0.5 Kg |
| Product Type | Cyanoacrylate Fuming Pans |
| Material | Aluminum |
| For Use With (Application) | Holding cyanoacrylate adhesive such as super glue |