missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Purifier™ Clean Bench IV Bar Kits

Catalog No. 16107180
Change view
Click to view available options
For Use With (Equipment):
3, 4 and 6 ft. Purifier Horizontal Clean Benches
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
16107180 3, 4 and 6 ft. Purifier Horizontal Clean Benches
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16107180 Supplier Labconco™ Supplier No. 3697500

Includes

Mounting hardware, four hangers and instructions for installation

Support intravenous solution bottles and bags

  • 3 ft. and 4 ft. Purifier Horizontal Clean Benches accommodate one IV bar
  • 6 ft. benches accommodate two bars

Specifications

For Use With (Equipment) 3, 4 and 6 ft. Purifier Horizontal Clean Benches
Weight (English) 5 lb.
Weight (Metric) 2.3 Kg
Product Type IV Bar Kit
For Use With (Application) 3, 4 and 6 ft. Purifier™ horizontal clean benches