missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Protector™ Standard Storage Cabinet

The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.

Supplier:  Labconco™ 9904000

 View more versions of this product

Catalog No. 16-305-67


Only null left

Includes:

Four leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30 in.


Description

Description

The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.

  • Support loads up to 800 lb. (363kg)
  • Durable epoxy-coated steel construction
  • Attractive glacier white exterior complements laboratory casework and Labconco fume hoods
  • Manual-closing, non-locking door(s)

Order Info

Work surface and interior shelf kits are sold separately

Specifications

Specifications

Storage Cabinet
Ideal for storage and venting of small laboratory instrument and mild acid and other reagent
24 in.
Right Hinge, Manual Close
55.88 cm
60.96 cm
1
800 lb.
Light Gray
22 in.
Four Leveling Feet and One 8 in. D Filler Panel to Increase Cabinet Depth from 22 to 30 in.
54.43 kg
120 lb.
362.8 kg
31.5 to 32.75 in.
ADA
80 to 83.18 cm
Epoxy-coated Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions